Learn More
Medchemexpress LLC HY-113008 500mg Medchemexpress, Urocanic acid CAS:104-98-3 Purity:>98%
SDP
Catalog No. 501969213
Encompass_Preferred
Medchemexpress, HY-113008 500mg Urocanic acid CAS:104-98-3 Urocanic acid, produced in the upper layers of mammalian skin, is a major absorber of ultraviolet radiation (UVR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9213 Supplier Medchemexpress LLC Supplier No. HY113008500MG
Medchemexpress, HY-113008 500mg Urocanic acid CAS:104-98-3 Urocanic acid, produced in the upper layers of mammalian skin, is a major absorber of ultraviolet radiation (UVR). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.