Learn More
Bachem Urocortin (human) Trifluoroacetate
SDP
Supplier:  Bachem 4027201.0500
Encompass_Preferred
Urocortin (human) Trifluoroacetate, H-Asp-Asn-Pro-Ser-Leu-Ser-Ile-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Thr-Leu-Leu-Glu-Leu-Ala-Arg-Thr-Gln-Ser-Gln-Arg-Glu-Arg-Ala-Glu-Gln-Asn-Arg-Ile-Ile-Phe-Asp-Ser-Val-NH2, 176591-49-4, C204H337N63O64, Mr 4696.31, 0.5mg. Using a genomic library, human urocortin (UCN) has been cloned and localized to human chromosome 2. Human and rat urocortin share 95% identity within the mature peptide region. Human urocortin binds with high affinity to CRF receptor types 1, 2a, and 2b, stimulates cAMP accumulation from cells transfected with these receptors and acts in vitro to release ACTH from dispersed rat anterior pituitary cells.
Catalog No. 50-259-768