Learn More
CPC Scientific H-Ile-Val-Leu-Ser-Leu-Asp-Val-Pro-Ile-Gly-Leu-Leu-Gln-Ile-Leu-Leu-Glu-Gln-Ala-Arg-Ala-Arg-Ala-Ala-Arg-Glu-Gln-Ala-Thr-Thr-Asn-Ala-Arg-Ile-Leu-Ala-Arg-Val-Gly-His-Cys-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific UROC003B
Encompass
As a new member of the CRF family of neuropeptides, this peptide selectively binds to the type 2 CRF receptor (CRF-R2) while exhibiting no appreciable activity on CRF-R1.ONE-LETTER SEQUENCE: IVLSLDVPIGLLQILLEQARARAAREQATTNARILARVGHC-NH2MOLECULAR FORMULA: C194H339N63O54S1MOLECULAR WEIGHT:4450.28STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [398001-88-2], RESEARCH AREA: NeuropeptidesREFERENCES: K. Lewis et al., Proc. Natl. Acad. Sci. USA , 98, 7570 (2001); T. M.Reyes et al., Proc. Natl. Acad. Sci. USA, 98, 2843 (2001)
Catalog No. 50-211-0591