Learn More
Bachem Urocortin II (mouse) Trifluoroacetate
SDP
Supplier:  Bachem 4048157.0500
Encompass_Preferred
Urocortin II (mouse) Trifluoroacetate, H-Val-Ile-Leu-Ser-Leu-Asp-Val-Pro-Ile-Gly-Leu-Leu-Arg-Ile-Leu-Leu-Glu-Gln-Ala-Arg-Tyr-Lys-Ala-Ala-Arg-Asn-Gln-Ala-Ala-Thr-Asn-Ala-Gln-Ile-Leu-Ala-His-Val-NH2, 330648-32-3, C187H320N56O50, Mr 4152.95, 0.5mg. Ucn II (mouse). Urocortin II (mouse) exhibited considerably higher affinity for both splice variants of the type 2 CRF receptor (CRF-R2) than for CRF-R1. The potencies of Ucn II (mouse) in activating CRF-R2a and CRF-R2b were nearly equivalent to that of urocortin (rat).
Catalog No. NC1407024