Learn More
CPC Scientific H-Phe-Thr-Leu-Ser-Leu-Asp-Val-Pro-Thr-Asn-Ile-Met-Asn-Ile-Leu-Phe-Asn-Ile-Asp-Lys-Ala-Lys-Asn-Leu-Arg-Ala-Lys-Ala-Ala-Ala-Asn-Ala-Gln-Leu-Met-Ala-Gln-Ile-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific UROC007B
Encompass
ONE-LETTER SEQUENCE: FTLSLDVPTNIMNILFNIDKAKNLRAKAAANAQLMAQI-NH2MOLECULAR FORMULA: C186H312N52O52S2MOLECULAR WEIGHT:4173STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [357952-10-4], RESEARCH AREA: NeuropeptidesREFERENCES: K.Lewis et al., Proc. Natl. Acad. Sci. USA , 98, 7570 (2001)
Catalog No. 50-211-0597