Learn More
CPC Scientific H-Asp-Asp-Pro-Pro-Leu-Ser-Ile-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Thr-Leu-Leu-Glu-Leu-Ala-Arg-Thr-Gln-Ser-Gln-Arg-Glu-Arg-Ala-Glu-Gln-Asn-Arg-Ile-Ile-Phe-Asp-Ser-Val-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific UROC002A
Encompass
Peptide that is more potent than CRF at binding and activating type-2 CRF receptors and shares ~95% sequence with human version. As a mammalian neuropeptide related to fish urotensin and corticotropin-releasing factor, it also serves as an inducer of c-Fos in regions enriched in type-2 CRF receptors.ONE-LETTER SEQUENCE: DDPPLSIDLTFHLLRTLLELARTQSQRERAEQNRIIFDSV-NH2MOLECULAR FORMULA: C206H338N62O64MOLECULAR WEIGHT:4707.33STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [171543-83-2], RESEARCH AREA: NeuropeptidesREFERENCES: J. Vaughan et al, Nature, 378, 287(1995)
Catalog No. 50-211-0588