Learn More
Aviva Systems Biology Corp UROD Antibody - N-terminal region (ARP41724_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41724_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against UROD. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human UROD. Alternate names: PCT, UPDApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-57968