Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Urofix Prefills
SDP

Catalog No. 22031888
Change view
Click to view available options
Quantity:
120 mL x 96 Vials/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-031-888 120 mL x 96 Vials/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-031-888 Supplier StatLab Supplier No. UF08120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An Excellent Fixative for Prostate Biopsies

  • Preserves nuclear detail extremely well, and penetrates tissue quicker, thereby shortening fixation times
  • 120 mL Prefills are filled 50% with fixative (60 mL)
  • 96 Vials/Cs.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Quantity 120 mL x 96 Vials/Cs.
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.