Learn More
Sino Biological Urokinase / PLAU Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10815MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human uPA / PLAU (rhPLAU; Catalog#10815-H08H; Met 1-Leu 431; NP_002649.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-7608