Learn More
Sino Biological Urokinase / PLAU Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10815T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Urokinase / PLAU (rh Urokinase / PLAU; Catalog#10815-H08H; NP_002649.1; Met1-Leu431). Urokinase / PLAU specific IgG was purified by Human Urokinase / PLAU affinity chromatography.
Catalog No. 50-224-7468