Learn More
Selleck Chemical LLC Urolithin A 10mg 1143-70-0 3,8-Dihydroxy Urolithin; 2',7-Dihydroxy-3,4-benzocoumarin
SDP
Supplier:  Selleck Chemical LLC S531210MG
Encompass_Preferred
Urolithin A (3,8-Dihydroxy Urolithin, 2',7-Dihydroxy-3,4-benzocoumarin), a metabolite of ellagitannin, is a first-in-class natural compound that induces mitophagy both in vitro and in vivo following oral consumption. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8185