Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Urolithin A | 1143-70-0 | MFCD20275235 | 228.20 g/mol | C13H8O4 | 1 ML
SDP

Catalog No. 5000213898
Encompass_Preferred

Urolithin A is a gut-microbial metabolite used as a research reagent to study mitophagy, autophagy, apoptosis, and cellular metabolism. The product is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and includes preparation guidance and storage recommendations for experimental use.

  • Suitable for mitophagy, autophagy, and apoptosis studies.
  • Ready-to-use 10 mM solution in DMSO for immediate experimental use.
  • Molecular formula C13H8O4; molecular weight 228.20 g/mol.
  • Datasheet includes solvent volumes for preparing 10 mM stocks.
  • Recommended storage conditions provided for powder and in-solution stability.
  • Intended for research use only.

Catalog No. 50-002-13898 Supplier Medchemexpress LLC Supplier No. HY10059910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Urolithin A is a gut-microbial metabolite used as a research reagent to study mitophagy, autophagy, apoptosis, and cellular metabolism. The product is provided as a ready-to-use 10 mM solution in DMSO (1 mL) and includes preparation guidance and storage recommendations for experimental use.

  • Suitable for mitophagy, autophagy, and apoptosis studies.
  • Ready-to-use 10 mM solution in DMSO for immediate experimental use.
  • Molecular formula C13H8O4; molecular weight 228.20 g/mol.
  • Datasheet includes solvent volumes for preparing 10 mM stocks.
  • Recommended storage conditions provided for powder and in-solution stability.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.