Learn More
Selleck Chemical LLC Urolithin A S5312-10mM/1mL
SDP
Catalog No. 503112027
Encompass_Preferred
Urolithin A (3 8-Dihydroxy Urolithin 2 7-Dihydroxy-3 4-benzocoumarin) a metabolite of ellagitannin is a first-in-class natural compound that induces mitophagy both in vitro and in vivo following oral consumption
Catalog No. 50-311-2027 Supplier Selleck Chemical LLC Supplier No. S531210MM/1ML
Add to Cart
Add to Cart
Urolithin A (3 8-Dihydroxy Urolithin 2 7-Dihydroxy-3 4-benzocoumarin) a metabolite of ellagitannin is a first-in-class natural compound that induces mitophagy both in vitro and in vivo following oral consumption