Learn More
TARGETMOL CHEMICALS INC UROLITHIN A 25MG
SDP
Catalog No. 502576342
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Urolithin A is a secondary metabolite of ellagic acid a polyphenolic antioxidant that has antiproliferative anti-inflammatory and anti-oxidant properties. purity: 99%
Catalog No. 50-257-6342 Supplier TARGETMOL CHEMICALS INC Supplier No. T717425MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Urolithin A is a secondary metabolite of ellagic acid a polyphenolic antioxidant that has antiproliferative anti-inflammatory and anti-oxidant properties. purity: 99%