Learn More
Medchemexpress LLC HY-100599 100mg , Urolithin A CAS:1143-70-0 Purity:98%
SDP
Catalog No. 501493007
Encompass_Preferred
Medchemexpress, HY-100599 100mg Urolithin A CAS:1143-70-0 Formula:C13H8O4 IC50: 43.9 μM (T24 cell) , 49 μM (Caco-2cell) Purity:98% Urolithin A is an intestinal metabolite of ellagic acid with antioxidant and antiproliferative effects; inhibits T24 and Caco-2 cell growth with IC 50 values of 43.9 and 49 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3007 Supplier Medchemexpress LLC Supplier No. HY100599100MG
Add to Cart
Add to Cart
Medchemexpress, HY-100599 100mg Urolithin A CAS:1143-70-0 Formula:C13H8O4 IC50: 43.9 μM (T24 cell) , 49 μM (Caco-2cell) Purity:98% Urolithin A is an intestinal metabolite of ellagic acid with antioxidant and antiproliferative effects; inhibits T24 and Caco-2 cell growth with IC 50 values of 43.9 and 49 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.