Learn More
TARGETMOL CHEMICALS INC UROLITHIN B 50MG
SDP
Catalog No. 502577401
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Urolithin B is one of the gut microbial metabolites of ellagitannins and is found in diverse plant foods including pomegranates berries walnuts tropical fruits and medicinal herbs. Urolithin B inhibits NF-(kappa)B activity by reducing the phosphorylation and degradation of I(kappa)B(alpha) and suppresses the phosphorylation of JNK ERK and Akt and enhances the phosphorylation of AMPK.Urolithin B is also a regulator of skeletal muscle mass. Urolithin B has anti-inflammatory and antioxidant effects. purity: 99%
Catalog No. 50-257-7401 Supplier TARGETMOL CHEMICALS INC Supplier No. T1326550MG
Also available in 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Urolithin B is one of the gut microbial metabolites of ellagitannins and is found in diverse plant foods including pomegranates berries walnuts tropical fruits and medicinal herbs. Urolithin B inhibits NF-(kappa)B activity by reducing the phosphorylation and degradation of I(kappa)B(alpha) and suppresses the phosphorylation of JNK ERK and Akt and enhances the phosphorylation of AMPK.Urolithin B is also a regulator of skeletal muscle mass. Urolithin B has anti-inflammatory and antioxidant effects. purity: 99%