Learn More
Medchemexpress LLC Urolithin D (3,4,8,9-tetrahydroxy urolithin) | 131086-98-1 | MFCD00954508 | 99.7% | 260.20 | C13H8O6 | 5 MG
SDP
Catalog No. 5000262792
Encompass_Preferred
Urolithin D (3,4,8,9-tetrahydroxy urolithin) is a research-grade small molecule used as an analytical standard and biochemical reagent. It is supplied in solid and solution formats with a high reported purity and accompanying documentation to support research, analytical, and signaling pathway studies.
  • High purity analytical standard (99.72%).
  • Supplied in small solid quantities suitable for analytical work, including 5 mg packs.
  • Available as a DMSO solution for immediate experimental use (10 mM, 1 mL).
  • Datasheet, certificate of analysis, and SDS provided for quality and safety verification.
  • Suitable for studies of EphA/ephrin signaling and related biochemical pathways.
Catalog No. 50-002-62792 Supplier Medchemexpress LLC Supplier No. HY1331785MG
Urolithin D (3,4,8,9-tetrahydroxy urolithin) is a research-grade small molecule used as an analytical standard and biochemical reagent. It is supplied in solid and solution formats with a high reported purity and accompanying documentation to support research, analytical, and signaling pathway studies.
  • High purity analytical standard (99.72%).
  • Supplied in small solid quantities suitable for analytical work, including 5 mg packs.
  • Available as a DMSO solution for immediate experimental use (10 mM, 1 mL).
  • Datasheet, certificate of analysis, and SDS provided for quality and safety verification.
  • Suitable for studies of EphA/ephrin signaling and related biochemical pathways.