Learn More
Medchemexpress LLC Urolithin M5 | 91485-02-8 | 5 MG
SDP
Catalog No. 5000417708
Encompass_Preferred
Urolithin M5 is an orally active influenza virus neuraminidase inhibitor and neuroprotective agent. It inhibits viral replication, including oseltamivir-resistant strains, and offers protection in infected mammals by reducing lung edema. It also provides neuroprotection by reducing reactive oxygen species, inhibiting neuronal apoptosis, and promoting neurite outgrowth.
  • Orally active influenza virus neuraminidase inhibitor
  • Neuroprotective agent
  • Suppresses viral neuraminidase activity
  • Inhibits influenza virus replication
  • Protects infected mammals and improves lung edema
  • Reduces reactive oxygen species production
  • Inhibits neuronal apoptosis
  • Restores mitochondrial transmembrane potential
  • Promotes neurite outgrowth in damaged neuronal cells
  • Suitable for research related to influenza and Alzheimer's disease
Catalog No. 50-004-17708 Supplier Medchemexpress LLC Supplier No. HYN79225MG
Urolithin M5 is an orally active influenza virus neuraminidase inhibitor and neuroprotective agent. It inhibits viral replication, including oseltamivir-resistant strains, and offers protection in infected mammals by reducing lung edema. It also provides neuroprotection by reducing reactive oxygen species, inhibiting neuronal apoptosis, and promoting neurite outgrowth.
  • Orally active influenza virus neuraminidase inhibitor
  • Neuroprotective agent
  • Suppresses viral neuraminidase activity
  • Inhibits influenza virus replication
  • Protects infected mammals and improves lung edema
  • Reduces reactive oxygen species production
  • Inhibits neuronal apoptosis
  • Restores mitochondrial transmembrane potential
  • Promotes neurite outgrowth in damaged neuronal cells
  • Suitable for research related to influenza and Alzheimer's disease