Learn More
Diagnostic Biosystems UROPLAKIN 11 UPK2 Uroplakin II
Catalog No. 502407427
Encompass_Preferred
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. Uroplakins are markers of terminally differentiated urothelium. Uroplakin II (UPII) is a newly described sensitive marker for urothelial carcinoma (UC). The expression profile of UPII in different types of UC and its utility in the diagnostic setting are needed.
Catalog No. 50-240-7427 Supplier Diagnostic Biosystems Supplier No. PDR177
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. Uroplakins are markers of terminally differentiated urothelium. Uroplakin II (UPII) is a newly described sensitive marker for urothelial carcinoma (UC). The expression profile of UPII in different types of UC and its utility in the diagnostic setting are needed.