Learn More
Medchemexpress LLC HY-P1542B 500ug Medchemexpress, Urotensin I (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1542B500UG
Encompass_Preferred
Medchemexpress, HY-P1542B 500ug Urotensin I (TFA) CAS: Urotensin I (Catostomus urotensin I) TFA, a CRF-like neuropeptide, acts as an agonist of CRF receptor with pEC50s of 11.46, 9.36 and 9.85 for human CRF1, human CRF2 and rat CRF2α receptors in CHO cells, and Kis of 0.4, 1.8, and 5.7 nM for hCRF1, rCRF2α and mCRF2β receptors, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3066