Learn More
Medchemexpress LLC HY-P1164A 1mg Medchemexpress, Urotensin II (114-124), human (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1164A1MG
Encompass_Preferred
Medchemexpress, HY-P1164A 1mg Urotensin II (114-124), human (TFA) CAS: Urotensin II (114-124), human TFA, an 11-amino acid residue peptide, is a potent vasoconstrictor and agonist for the orphan receptor GPR14 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4158
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.