Learn More
CPC Scientific H-Glu-Thr-Pro-Asp-Cys-Phe-Trp-Lys-Tyr-Cys-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific UROT003B
Encompass
A vasoactive "somatostatin-like" cyclic peptide originally derived from fish spinal cords that has recently been cloned from man. It was found to be the most potent mammalian vasoconstrictor identified so far and may have additional activities, as urotensin II immunoreactivity was also found within the central nervous system and endocrine tissues.ONE-LETTER SEQUENCE: ETPDCFWKYCV (Cys5 and 10 bridge)MOLECULAR FORMULA: C64H85N13O18S2MOLECULAR WEIGHT:1388.59STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [251293-28-4], SYNONYMS: U-II (human), Urotensin II (human)RESEARCH AREA: CardiovascularREFERENCES: K.L.Ong et al., Cardiovasc. Drugs Ther., 19, 65 (2005); R.S.Ames et al., Nature, 401, 282 (1999); Y.Coulouarn et al., Proc. Natl. Acad. Sci. USA, 95, 15803 (1998)
Catalog No. 50-211-0602