Learn More
Cayman Chemical Ursocholic acid 10mg
SDP
Catalog No. 502992443
Encompass_Preferred
A bile acid; dietary administration decreases gallbladder cholesterol, phospholipid, and bile acid levels and the occurrence of gallstones in male mice fed a lithogenic diet at 0.25%
Catalog No. 50-299-2443 Supplier Cayman Chemical Supplier No. 2988310 MG
A bile acid; dietary administration decreases gallbladder cholesterol, phospholipid, and bile acid levels and the occurrence of gallstones in male mice fed a lithogenic diet at 0.25%