Learn More
Medchemexpress LLC Ursocholic acid | 2955-27-3 | 99.9% | 408.57 | 50 MG
SDP
Catalog No. 5000435819
Encompass_Preferred
Ursocholic acid is a bile acid found in mammalian bile that is converted to deoxycholic acid (UDC) by mouse intestinal flora. It functions as a gallstone dissolving agent in the liver by anti-apoptosis, anti-inflammatory, immunomodulatory, bile regulation, and coordinated changes in mitochondrial integrity and cell signaling. It also has favorable effects on bones in patients with chronic cholestasis.
  • Acts as a gallstone dissolving agent
  • Exhibits anti-apoptosis and anti-inflammatory properties
  • Modulates immune responses
  • Regulates bile and mitochondrial integrity
  • Shows beneficial effects on bones in chronic cholestasis
  • Increases osteoblast differentiation and mineralization
  • Reduces inflammatory cytokine levels and intestinal villus damage in rats
Catalog No. 50-004-35819 Supplier Medchemexpress LLC Supplier No. HY11321250MG
Ursocholic acid is a bile acid found in mammalian bile that is converted to deoxycholic acid (UDC) by mouse intestinal flora. It functions as a gallstone dissolving agent in the liver by anti-apoptosis, anti-inflammatory, immunomodulatory, bile regulation, and coordinated changes in mitochondrial integrity and cell signaling. It also has favorable effects on bones in patients with chronic cholestasis.
  • Acts as a gallstone dissolving agent
  • Exhibits anti-apoptosis and anti-inflammatory properties
  • Modulates immune responses
  • Regulates bile and mitochondrial integrity
  • Shows beneficial effects on bones in chronic cholestasis
  • Increases osteoblast differentiation and mineralization
  • Reduces inflammatory cytokine levels and intestinal villus damage in rats