Learn More
Cayman Chemical Ursocholic Acid-d4, Purity: ≥99% deuterated forms (d1-d4), 10 mg
SDP
Catalog No. 502909930
Encompass_Preferred
An internal standard for the quantification of ursocholic acid by GC- or LC-MS
Catalog No. 50-290-9930 Supplier Cayman Chemical Supplier No. 3541910 MG
An internal standard for the quantification of ursocholic acid by GC- or LC-MS