Learn More
Medchemexpress LLC Ursodeoxycholic acid | 128-13-2 | 99.8% | 392.57 | 250 MG
SDP
Catalog No. 5000348248
Encompass_Preferred
Ursodeoxycholic acid is a secondary bile acid formed from the transformation of (cheno)deoxycholic acid by intestinal bacteria. It acts as a key regulator of intestinal barrier integrity and is essential for lipid metabolism. It functions as a signaling molecule by interacting with bile acid-activated receptors, including G-protein coupled bile acid receptor 5 (TGR5, GPCR19) and the farnesoid X receptor (FXR). It is used for research into various hepatic and gastrointestinal diseases, and also reduces ACE2 expression, which is beneficial for reducing SARS-CoV-2 infection. It is orally active.
  • Regulates intestinal barrier integrity
  • Essential for lipid metabolism
  • Interacts with bile acid-activated receptors (TGR5, GPCR19, FXR)
  • Used for research on hepatic and gastrointestinal diseases
  • Reduces ACE2 expression, beneficial for reducing SARS-CoV-2 infection
  • Orally active
Catalog No. 50-003-48248 Supplier Medchemexpress LLC Supplier No. 250MGHY13771R
Ursodeoxycholic acid is a secondary bile acid formed from the transformation of (cheno)deoxycholic acid by intestinal bacteria. It acts as a key regulator of intestinal barrier integrity and is essential for lipid metabolism. It functions as a signaling molecule by interacting with bile acid-activated receptors, including G-protein coupled bile acid receptor 5 (TGR5, GPCR19) and the farnesoid X receptor (FXR). It is used for research into various hepatic and gastrointestinal diseases, and also reduces ACE2 expression, which is beneficial for reducing SARS-CoV-2 infection. It is orally active.
  • Regulates intestinal barrier integrity
  • Essential for lipid metabolism
  • Interacts with bile acid-activated receptors (TGR5, GPCR19, FXR)
  • Used for research on hepatic and gastrointestinal diseases
  • Reduces ACE2 expression, beneficial for reducing SARS-CoV-2 infection
  • Orally active