Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ursodeoxycholic acid | 128-13-2 | 99.8% | 392.57 | 5 G
SDP

Catalog No. 5000349617
Encompass_Preferred

Ursodeoxycholic acid is a reference standard with a molecular weight of 392.57. It has a high purity of 99.8% (HPLC) and is supplied in a brown glass bottle.

  • Reference standard
  • High purity of 99.8% (HPLC)
  • Packaged in a brown glass bottle
  • Stored at -20°C, sealed, away from light and moisture

Catalog No. 50-003-49617 Supplier Medchemexpress LLC Supplier No. HY137715G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ursodeoxycholic acid is a reference standard with a molecular weight of 392.57. It has a high purity of 99.8% (HPLC) and is supplied in a brown glass bottle.

  • Reference standard
  • High purity of 99.8% (HPLC)
  • Packaged in a brown glass bottle
  • Stored at -20°C, sealed, away from light and moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.