Learn More
Medchemexpress LLC Ursodeoxycholic acid | 128-13-2 | 99.94% | 392.57 | 5 G
SDP
Catalog No. 5000350579
Encompass_Preferred
Ursodeoxycholic acid (Ursodeoxycholate) is a secondary bile acid that regulates intestinal barrier integrity and is essential for lipid metabolism. It functions as a signaling molecule by interacting with bile acid-activated receptors, including TGR5 and FXR. This compound is used in research for hepatic and gastrointestinal diseases, and reduces ACE2 expression, which is beneficial for reducing SARS-CoV-2 infection. It is orally active.
  • Secondary bile acid
  • Regulates intestinal barrier integrity
  • Essential for lipid metabolism
  • Interacts with TGR5 and FXR receptors
  • Used in hepatic and gastrointestinal disease research
  • Reduces ACE2 expression
  • Orally active
Catalog No. 50-003-50579 Supplier Medchemexpress LLC Supplier No. HY13771A5G
Add to Cart
Add to Cart
Ursodeoxycholic acid (Ursodeoxycholate) is a secondary bile acid that regulates intestinal barrier integrity and is essential for lipid metabolism. It functions as a signaling molecule by interacting with bile acid-activated receptors, including TGR5 and FXR. This compound is used in research for hepatic and gastrointestinal diseases, and reduces ACE2 expression, which is beneficial for reducing SARS-CoV-2 infection. It is orally active.
  • Secondary bile acid
  • Regulates intestinal barrier integrity
  • Essential for lipid metabolism
  • Interacts with TGR5 and FXR receptors
  • Used in hepatic and gastrointestinal disease research
  • Reduces ACE2 expression
  • Orally active