Learn More
Medchemexpress LLC Ursodeoxycholic acid | 128-13-2 | MFCD00003680 | >99.9% | 392.57 | 1 ML
SDP
Catalog No. 5000351440
Encompass_Preferred
Ursodeoxycholic acid (UDCA), also known as Ursodiol, is a naturally occurring secondary bile acid. It is formed in the intestines from the transformation of (cheno)deoxycholic acid by intestinal bacteria. UDCA plays a crucial role as a key regulator of lipid and glucose metabolism. This product is a 10 mM solution in 1 mL of DMSO, offering a purity of 99.94%.
  • Naturally occurring secondary bile acid.
  • Regulator of lipid and glucose metabolism.
  • Used to dissolve cholesterol gallstones.
  • Treats cholestatic forms of liver disease.
  • Potential therapeutic agent for inflammatory bowel disease.
  • Potential therapeutic agent for diabetes and other metabolic disorders.
  • Supplied as a 10 mM solution in DMSO.
Catalog No. 50-003-51440 Supplier Medchemexpress LLC Supplier No. HY13771A10MM/1ML
Add to Cart
Add to Cart
Ursodeoxycholic acid (UDCA), also known as Ursodiol, is a naturally occurring secondary bile acid. It is formed in the intestines from the transformation of (cheno)deoxycholic acid by intestinal bacteria. UDCA plays a crucial role as a key regulator of lipid and glucose metabolism. This product is a 10 mM solution in 1 mL of DMSO, offering a purity of 99.94%.
  • Naturally occurring secondary bile acid.
  • Regulator of lipid and glucose metabolism.
  • Used to dissolve cholesterol gallstones.
  • Treats cholestatic forms of liver disease.
  • Potential therapeutic agent for inflammatory bowel disease.
  • Potential therapeutic agent for diabetes and other metabolic disorders.
  • Supplied as a 10 mM solution in DMSO.