Learn More
AdipoGen Ursodeoxycholic acid
SDP
Supplier:  AdipoGen CDXU0019G025
Encompass_Preferred
Chemical. CAS 128-13-2. Formula C24H40O4. MW 392.57. Synthetic. Endogenous hydrophilic bile acid. Antioxidant. Cytoprotective against oxidative stress and cell death. Hepatoprotective at cellular and molecular level, including stabilization of membranes. Antiapoptotic and antinecrotic. Targets the mitochondrial function and integrity, reduction of endoplasmatic stress and interactions with survival signals in cAMP, Akt, NF-kappaB, MAPK and PI3K signaling pathways. Chemopreventive against colorectal cancer by countering the tumor-promoting effects of secondary bile acids. Shows also effects on epidermal growth factor receptor EGFR signaling and COX-2 expression. Immunomodulator and anti-inflammatory compound. Modifies TLR4 and TLR9 signaling pathways and downregulates the production of proinflammatory tumor necrosis factor-alpha TNF-alpha. Pregnane X receptor agonist. Neuroprotective. Anticholestatic agent.
Catalog No. 50-255-9924