Learn More
Cayman Chemical Ursodeoxycholic Acid, Purity: ≥95%, CAS: 128-13-2, 5 g
SDP
Catalog No. 502988383
Encompass_Preferred
A secondary bile acid; formed via epimerization of CDCA; a metabolite of LCA in human liver microsomes; inhibits taurocholic acid uptake in HeLa cells expressing recombinant human NTCP (IC50 = 3.6 μM); inhibits apoptosis induced by DCA or ethanol in primary rat hepatocytes at 50 μM; dietary administration blocks DCA-induced increases in the number of TUNEL-positive hepatocytes in rats
Catalog No. 50-298-8383 Supplier Cayman Chemical Supplier No. 310165 G
A secondary bile acid; formed via epimerization of CDCA; a metabolite of LCA in human liver microsomes; inhibits taurocholic acid uptake in HeLa cells expressing recombinant human NTCP (IC50 = 3.6 μM); inhibits apoptosis induced by DCA or ethanol in primary rat hepatocytes at 50 μM; dietary administration blocks DCA-induced increases in the number of TUNEL-positive hepatocytes in rats