Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ursodeoxycholic acid, 99%, Thermo Scientific Chemicals

Catalog No. AAB2049003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2049003 1 g
AAB2049006 5 g
AAB2049014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2049003 Supplier Thermo Scientific Chemicals Supplier No. B2049003
  • $55.80
    $47.65 / Each of 1

    Save $8.15

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

A secondary bile acid

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-13-2
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003680
InChI Key RUDATBOHQWOJDD-UZVSRGJWSA-N
Synonym ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid
PubChem CID 31401
ChEBI CHEBI:9907
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Melting Point 201°C to 204°C
Quantity 1 g
Merck Index 14,9889
Formula Weight 392.56
Percent Purity 99%
Chemical Name or Material Ursodeoxycholic acid
EINECSNumber 204-879-3
RTECSNumber FZ2000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.