Learn More
Medchemexpress LLC Ursodeoxycholic acid-d4 | 347841-46-7 | 99.5% | 396.60 | 1 MG
SDP
Catalog No. 5000438407
Encompass_Preferred
Ursodeoxycholic acid-2,2,4,4-d4 is a deuterium-labeled form of Ursodeoxycholic acid (UDCA), a secondary bile acid. It plays a role in regulating intestinal barrier integrity and is crucial for lipid metabolism. It functions as a signaling molecule by interacting with bile acid-activated receptors, including G-protein coupled bile acid receptor 5 (TGR5, GPCR19) and the farnesoid X receptor (FXR).
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Beneficial for reducing SARS-CoV-2 infection by reducing ACE2 expression
  • Research on various hepatic and gastrointestinal diseases
  • Stable heavy isotopes for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • Appearance: Solid, white to off-white
Catalog No. 50-004-38407 Supplier Medchemexpress LLC Supplier No. HY113478S1MG
Ursodeoxycholic acid-2,2,4,4-d4 is a deuterium-labeled form of Ursodeoxycholic acid (UDCA), a secondary bile acid. It plays a role in regulating intestinal barrier integrity and is crucial for lipid metabolism. It functions as a signaling molecule by interacting with bile acid-activated receptors, including G-protein coupled bile acid receptor 5 (TGR5, GPCR19) and the farnesoid X receptor (FXR).
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Beneficial for reducing SARS-CoV-2 infection by reducing ACE2 expression
  • Research on various hepatic and gastrointestinal diseases
  • Stable heavy isotopes for quantitation during drug development
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
  • Appearance: Solid, white to off-white