Learn More
Cayman Chemical UrsodeoxycholIc AcId-d4 500ug
SDP
Catalog No. 502985713
Encompass_Preferred
An internal standard for the quantification of ursodeoxycholic acid by GC- or LC-MS
Catalog No. 50-298-5713 Supplier Cayman Chemical Supplier No. 21892500 UG
An internal standard for the quantification of ursodeoxycholic acid by GC- or LC-MS