Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ursodesoxycholic Acid, MP Biomedicals

Catalog No. ICN219738 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219738 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219738 Supplier MP Biomedicals, Inc Supplier No. 0521973801

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 128-13-2
Molecular Formula C24H40O4
Molecular Weight (g/mol) 392.58
MDL Number MFCD00003680
InChI Key RUDATBOHQWOJDD-UZVSRGJWSA-N
Synonym ursodeoxycholic acid, ursodiol, actigall, ursofalk, ursolvan, delursan, ursodeoxycholate, urso, urso forte, ursodesoxycholic acid
PubChem CID 31401
ChEBI CHEBI:9907
IUPAC Name (4R)-4-[(1R,3aS,3bR,4S,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
SMILES [H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Quantity 1 g
Formula Weight 392.29
Chemical Name or Material Ursodesoxycholic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.