Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ursolic Acid

Catalog No. U6753100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U6753100MG 100 g
U6753500MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U6753100MG Supplier Sigma Aldrich Supplier No. U6753100MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ursolic Acid

Specifications

CAS 121-33-5
Melting Point 81°C to 83°C
Boiling Point 170°C (15 mmHg, lit.)
Molecular Formula C8H8O3
Linear Formula 4-(HO)C6H3-3-(OCH3)CHO
MDL Number MFCD00006942
Quantity 100 g
Synonym 3beta-Hydroxy-12-ursen-28-ic acid
Molecular Weight (g/mol) 152.15 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.