Learn More
Enzo Life Sciences Ursolic acid (250mg). CAS: 77-52-1
SDP
Supplier:  Enzo Life Sciences BMLCT1050250
Encompass_Preferred
Inhibits endothelial cell proliferation and migration (IC50=5 µM) and angiogenesis. Purity: ≥90% (TLC). Solubility: Soluble in ethanol (5 mg/ml warm) or DMSO (>25 mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-0480