Learn More
TARGETMOL CHEMICALS INC Ursolic acid acetate 5MG
SDP
Catalog No. 502879038
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ursolic acid acetate (Acetylursolic acid) has cytotoxic activity, antimalarial activity, antitumor and anticancer activities. Acetylursolic acid has NO production inhibitory activities. Acetylursolic acid has inhibition against both acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) enzymes. Purity 98.3%
Catalog No. 50-287-9038 Supplier TARGETMOL CHEMICALS INC Supplier No. T38195MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Ursolic acid acetate (Acetylursolic acid) has cytotoxic activity, antimalarial activity, antitumor and anticancer activities. Acetylursolic acid has NO production inhibitory activities. Acetylursolic acid has inhibition against both acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) enzymes. Purity 98.3%