Learn More
Medchemexpress LLC Ursolic acid acetate | 7372-30-7 | 1 MG
SDP
Catalog No. 5000421424
Encompass_Preferred
Ursolic acid acetate (Acetylursolic acid) is a triterpenoid compound and an inhibitor of *Plasmodium falciparum* heat shock protein 90 (PfHsp90) with a KD of 8.16 μM. It is cytotoxic to KB cells, with an IC50 value of 8.4 μM. Ursolic acid acetate can be used in tumor and antimalarial research.
  • Triterpenoid compound
  • Inhibitor of Plasmodium falciparum heat shock protein 90 (PfHsp90) (KD = 8.16 μM)
  • Cytotoxic to KB cells (IC50 = 8.4 μM)
  • Potential for use in tumor and antimalarial research
  • Can reduce the ATPase activity of PfHsp90 in a dose-dependent manner (1-5 mM; 4.5 h)
  • Has an IC50 of 24.93 μM against the P. falciparum FcB1 strain
Catalog No. 50-004-21424 Supplier Medchemexpress LLC Supplier No. HYN28151MG
Ursolic acid acetate (Acetylursolic acid) is a triterpenoid compound and an inhibitor of *Plasmodium falciparum* heat shock protein 90 (PfHsp90) with a KD of 8.16 μM. It is cytotoxic to KB cells, with an IC50 value of 8.4 μM. Ursolic acid acetate can be used in tumor and antimalarial research.
  • Triterpenoid compound
  • Inhibitor of Plasmodium falciparum heat shock protein 90 (PfHsp90) (KD = 8.16 μM)
  • Cytotoxic to KB cells (IC50 = 8.4 μM)
  • Potential for use in tumor and antimalarial research
  • Can reduce the ATPase activity of PfHsp90 in a dose-dependent manner (1-5 mM; 4.5 h)
  • Has an IC50 of 24.93 μM against the P. falciparum FcB1 strain