Learn More
Medchemexpress LLC Ursolic acid acetate | 7372-30-7 | 5 MG
SDP
Catalog No. 5000418979
Encompass_Preferred
Ursolic acid acetate is a triterpenoid compound that acts as an inhibitor of Plasmodium falciparum heat shock protein 90 (PfHsp90) with a KD of 8.16 μM. It exhibits cytotoxicity to KB cells with an IC50 value of 8.4 μM. It has potential applications in tumor and antimalarial research. It can reduce the ATPase activity of PfHsp90 in a dose-dependent manner and shows an IC50 of 24.93 μM against the P. falciparum FcB1 strain.
  • Inhibitor of Plasmodium falciparum heat shock protein 90
  • Cytotoxic to KB cells with an IC50 value of 8.4 μM
  • Potential applications in tumor and antimalarial research
  • Reduces ATPase activity of PfHsp90 in a dose-dependent manner
  • Exhibits IC50 of 24.93 μM against P. falciparum FcB1 strain
Catalog No. 50-004-18979 Supplier Medchemexpress LLC Supplier No. HYN28155MG
Ursolic acid acetate is a triterpenoid compound that acts as an inhibitor of Plasmodium falciparum heat shock protein 90 (PfHsp90) with a KD of 8.16 μM. It exhibits cytotoxicity to KB cells with an IC50 value of 8.4 μM. It has potential applications in tumor and antimalarial research. It can reduce the ATPase activity of PfHsp90 in a dose-dependent manner and shows an IC50 of 24.93 μM against the P. falciparum FcB1 strain.
  • Inhibitor of Plasmodium falciparum heat shock protein 90
  • Cytotoxic to KB cells with an IC50 value of 8.4 μM
  • Potential applications in tumor and antimalarial research
  • Reduces ATPase activity of PfHsp90 in a dose-dependent manner
  • Exhibits IC50 of 24.93 μM against P. falciparum FcB1 strain