Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ursolic acid acetate | 7372-30-7 | MFCD09752431 | 98.0% | 498.74 g/mol | C32H50O4 | 10 MG
SDP

Catalog No. 5000289205
Encompass_Preferred

Ursolic acid acetate is an acetylated triterpenoid supplied as a research-grade solid for in vitro biochemical and cellular studies, including cancer and antimalarial research. It is provided at high purity and should be handled according to laboratory safety protocols.

  • Purity 98.0%.
  • Form solid, white to off-white.
  • CAS number 7372-30-7.
  • Molecular formula C32H50O4.
  • Molecular weight 498.74 g/mol.
  • Storage solid 4°C; in solvent -80°C (long-term) or -20°C (short-term); protect from light.
  • Intended for research use only; not for human or animal therapeutic use.

Catalog No. 50-002-89205 Supplier Medchemexpress LLC Supplier No. HYN281510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ursolic acid acetate is an acetylated triterpenoid supplied as a research-grade solid for in vitro biochemical and cellular studies, including cancer and antimalarial research. It is provided at high purity and should be handled according to laboratory safety protocols.

  • Purity 98.0%.
  • Form solid, white to off-white.
  • CAS number 7372-30-7.
  • Molecular formula C32H50O4.
  • Molecular weight 498.74 g/mol.
  • Storage solid 4°C; in solvent -80°C (long-term) or -20°C (short-term); protect from light.
  • Intended for research use only; not for human or animal therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.