Learn More
Indofine Chemical Ursolic Acid With Hpl, 10 Mg
SDP
Supplier:  Indofine Chemical 020037S 10 MG
Encompass_Preferred
URSOLIC ACID with HPLC 10 MG, Chromatography Standards, 3b-Hydroxyurs-12-en-28-oic acid, C30H48O3, MW: 456.71, 98% by HPLC, 77-52-1, MFCD00009621
Catalog No. 50-908-867