Learn More
AdipoGen Ursolic acid (1 g)
SDP
Supplier:  AdipoGen CDXU0400G001
Encompass_Preferred
Chemical. CAS: 77-52-1. Formula: C30H48O3. MW: 456.711. Ursolic acid is a pentacyclic triterpenoid that has been isolated from M. pumila. Ursolic acid has diverse biological effects, including anti-inflammatory, anticancer, antidiabetic, antiobesity, neuroprotective, cardioprotective, hepatoprotective, antioxidant and antiviral and antibacterial effects. Effects relevant to its anticancer properties include, apoptosis and autophagy induction, preventing angiogenesis, microtubule-destabilizing, induction of cell cycle arrest and inhibition of tumor growth. Ursolic acid inhibits NF-kappa signaling through suppression of IkappaBalpha kinase and p65 phosphorylation. It has been shown to enhance pancreatic beta-cell function in diabetic mice and activates the TGR5 receptor to enhance GLP-1 secretion. Its antiobesity potential is related to inhibiting pancreatic lipase and adipogenesis and increasing insulin sensitivity and inhibiting alpha-glucosidase.
Catalog No. 50-205-4680