Learn More
TARGETMOL CHEMICALS INC URSONIC ACID 100MG
SDP
Catalog No. 502580317
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ursonic Acid (TOS-BB-0966) belongs to the family of Ursane Triterpenes whose structure is based on the pentacyclic ursane skeleton. It has potential as HIV-1 Protease Inhibitor. purity: 99%
Catalog No. 50-258-0317 Supplier TARGETMOL CHEMICALS INC Supplier No. T3323100MG
Also available in 5 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ursonic Acid (TOS-BB-0966) belongs to the family of Ursane Triterpenes whose structure is based on the pentacyclic ursane skeleton. It has potential as HIV-1 Protease Inhibitor. purity: 99%