Learn More
Selleck Chemical LLC Ursonic acid S3902-1g
SDP
Catalog No. 503109077
Encompass_Preferred
Ursonic acid (Prunol Malol beta-Ursolic acid NSC4060 CCRIS 7123 TOS-BB-0966) present in many plants is a pentacyclic triterpenoid that can be used as a cosmetics additive and serve as a starting material for synthesis of more potent bioactive derivatives such as experimental antitumor agents Ursonic acid induces the apoptosis of human cancer cells through multiple signaling pathways
Catalog No. 50-310-9077 Supplier Selleck Chemical LLC Supplier No. S39021G
Ursonic acid (Prunol Malol beta-Ursolic acid NSC4060 CCRIS 7123 TOS-BB-0966) present in many plants is a pentacyclic triterpenoid that can be used as a cosmetics additive and serve as a starting material for synthesis of more potent bioactive derivatives such as experimental antitumor agents Ursonic acid induces the apoptosis of human cancer cells through multiple signaling pathways