Learn More
CPC Scientific H-Ile-Pro-Leu-Arg-Gly-Ala-Phe-Ile-Asn-Gly-Arg-Trp-Asp-Ser-Gln-Cys-His-Arg-Phe-Ser-Asn-Gly-Ala-Ile-Ala-Cys-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502108824
Encompass
Urumin is a host defense peptide isolated from the skin of the South Indian frog that shows antiviral activity (virucide) for H1 hemagglutinin-bearing human influenza A viruses. The peptide, that shows no homology to known peptide families, targets the H1 hemagglutinin protein of the conserved stalk region.SEQUENCE: H-Ile-Pro-Leu-Arg-Gly-Ala-Phe-Ile-Asn-Gly-Arg-Trp-Asp-Ser-Gln-Cys-His-Arg-Phe-Ser-Asn-Gly-Ala-Ile-Ala-Cys-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: IPLRGAFINGRWDSQCHRFSNGAIACAMOLECULAR FORMULA: C129H198N42O35S2MOLECULAR WEIGHT: 2961.35STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:Holthausen, David J., et al. "An Amphibian Host Defense Peptide Is Virucidal for Human H1 Hemagglutinin-Bearing Influenza Viruses." Immunity 46 (2017): 587-595.SKU(s): ATMP-009A, ATMP-009B
Catalog No. 50-210-8824 Supplier CPC Scientific Supplier No. ATMP009A
Urumin is a host defense peptide isolated from the skin of the South Indian frog that shows antiviral activity (virucide) for H1 hemagglutinin-bearing human influenza A viruses. The peptide, that shows no homology to known peptide families, targets the H1 hemagglutinin protein of the conserved stalk region.SEQUENCE: H-Ile-Pro-Leu-Arg-Gly-Ala-Phe-Ile-Asn-Gly-Arg-Trp-Asp-Ser-Gln-Cys-His-Arg-Phe-Ser-Asn-Gly-Ala-Ile-Ala-Cys-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: IPLRGAFINGRWDSQCHRFSNGAIACAMOLECULAR FORMULA: C129H198N42O35S2MOLECULAR WEIGHT: 2961.35STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:Holthausen, David J., et al. "An Amphibian Host Defense Peptide Is Virucidal for Human H1 Hemagglutinin-Bearing Influenza Viruses." Immunity 46 (2017): 587-595.SKU(s): ATMP-009A, ATMP-009B