Learn More
Cayman Chemical Urumin trifluoroacetate salt
SDP
Supplier:  Cayman Chemical 24277500 UG
Encompass_Preferred
An H1-selective influenza antiviral; inhibits H1 hemagglutinin-containing human influenza A viruses by 60 to >90%; intranasal administration of urumin (20 UG) prior to a lethal dose of PR8 influenza increases survival in mice
Catalog No. 50-293-4586