Learn More
MSC US Cargo Control Polyester Ratchet Strap Tie Down, 15 ft, 1 in, 400 lb Load Capacity, S-Hook
Catalog No. 5000528392
Encompass
This polyester ratchet strap tie down is designed for securing loads, offering a 400 lb load capacity and a 1200 lb breaking strength. It features a 15 ft length and 1 inch width, equipped with durable S-hooks for reliable attachment.
  • Ratchet strap tie down type
  • S-hook end type
  • 400 lb load capacity
  • Polyester tie down material
  • 15 ft tie down length
  • 1 in tie down width
  • Steel end material
  • Yellow color
  • 1200 lb breaking strength
  • 1 ft fixed end length
Catalog No. 50-005-28392 Supplier MSC Supplier No. 39105754
This polyester ratchet strap tie down is designed for securing loads, offering a 400 lb load capacity and a 1200 lb breaking strength. It features a 15 ft length and 1 inch width, equipped with durable S-hooks for reliable attachment.
  • Ratchet strap tie down type
  • S-hook end type
  • 400 lb load capacity
  • Polyester tie down material
  • 15 ft tie down length
  • 1 in tie down width
  • Steel end material
  • Yellow color
  • 1200 lb breaking strength
  • 1 ft fixed end length