Learn More
MSC US Cargo Control Shackle, Galvanized Steel, 4000 lb Working Load Limit, Bolt, Nut & Cotter Pin
Catalog No. 5000529216
Encompass
The US Cargo Control BTAS12 is a shackle designed with a bolt, nut, and cotter pin. It features a 4000 lb working load limit and is constructed from galvanized steel, making it suitable for various lifting and rigging applications.
  • Constructed from durable galvanized steel.
  • Features a secure bolt, nut, and cotter pin design.
  • Offers a 4000 lb working load limit.
  • Meets Federal Specification RR-C-271 & ASTM A148M standards.
  • Ideal for demanding lifting and rigging tasks.
Catalog No. 50-005-29216 Supplier MSC Supplier No. 14602429
The US Cargo Control BTAS12 is a shackle designed with a bolt, nut, and cotter pin. It features a 4000 lb working load limit and is constructed from galvanized steel, making it suitable for various lifting and rigging applications.
  • Constructed from durable galvanized steel.
  • Features a secure bolt, nut, and cotter pin design.
  • Offers a 4000 lb working load limit.
  • Meets Federal Specification RR-C-271 & ASTM A148M standards.
  • Ideal for demanding lifting and rigging tasks.