Learn More
MSC US Cargo Control Shackle, Screw Pin, 880 Lb, Stainless Steel, 5/16 in Pin
Catalog No. 5000528318
Encompass
This shackle is designed for various lifting and rigging applications, offering a reliable connection point. Constructed from durable stainless steel, it features a screw pin for secure attachment and has a working load limit of 880 Lb. It is suitable for tasks requiring a 5/16 in pin diameter.
  • Features a screw pin type.
  • Provides a load capacity of 880 Lb.
  • Made from stainless steel for durability.
  • Equipped with a 5/16 in pin diameter.
  • Has a jaw inside height of 1.1300 in.
  • Offers an inside width and jaw opening of 0.4700 in.
  • Conforms to ASTM A148M standards.
Catalog No. 50-005-28318 Supplier MSC Supplier No. 38239737
This shackle is designed for various lifting and rigging applications, offering a reliable connection point. Constructed from durable stainless steel, it features a screw pin for secure attachment and has a working load limit of 880 Lb. It is suitable for tasks requiring a 5/16 in pin diameter.
  • Features a screw pin type.
  • Provides a load capacity of 880 Lb.
  • Made from stainless steel for durability.
  • Equipped with a 5/16 in pin diameter.
  • Has a jaw inside height of 1.1300 in.
  • Offers an inside width and jaw opening of 0.4700 in.
  • Conforms to ASTM A148M standards.